• USA Home
  • Search Results

  • hydrogenhexachloroiridateivhydrate40695anhydrousbasis11080284111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrogen hexachloroiridate(IV) hydrate

2 Products
Synonym:   Dihydrogen hexachloroiridate(IV) hydrate, Hexachloroiridium(IV) acid hydrate, Hydrogen iridium hexachloride
Image of Hydrogen hexachloroiridate(IV) hydrate
CAS Number:
Linear Formula:
 H2Cl6Ir · xH2O
Molecular Weight:
 406.95 (anhydrous basis)
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More