• USA Home
  • Search Results

  • hydrogenperoxidesolution3401772284111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydrogen peroxide solution

14 Products
Image of Hydrogen peroxide solution
CAS Number:
Empirical Formula (Hill Notation):
 H2O2
Molecular Weight:
 34.01
 Beilstein/REAXYS Number:
 3587191
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Restriction
  Refractive Index
  Greener Alternative Principles
  Physical Form
  Suitability
  Color
  Media Suitability
  Solubility
  Melting Point
  Vapor density vs air
  Quality clarifier
  Purity Grade
  Shelf Life
  Additive
  Density
  Concentration
  Assay
Show More

Product #
Description
216763 Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent, Green Alternative
H3410 Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications,
H1009 Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer,
316989 Hydrogen peroxide solution, contains potassium stannate as inhibitor, 30-32 wt. % in water, semiconductor grade, 99.999% trace metals basis,
516813 Hydrogen peroxide solution, 50 wt. % in H2O, stabilized,
16911 Hydrogen peroxide solution, for ultratrace analysis,
95321 Hydrogen peroxide solution, ≥30%, for trace analysis,
31642 Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.,
323381 Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O,
349887 Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O,
88597 Hydrogen peroxide solution, 3%, for microbiology,
18304 Hydrogen peroxide solution, 34.5-36.5%,
95299 Hydrogen peroxide solution, purum p.a., ≥35% (RT),
95294 Hydrogen peroxide solution, tested according to Ph Eur,