• USA Home
  • Search Results

  • hydroquinidine32643143555811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroquinidine

1 Products
Synonym:   (9S)-10,11-Dihydro-6′-methoxycinchonan-9-ol, Dihydroquinidine
Image of Hydroquinidine
CAS Number:
Empirical Formula (Hill Notation):
 C20H26N2O2
Molecular Weight:
 326.43
 Beilstein/REAXYS Number:
 91441
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
359343 Hydroquinidine, 95%,