• USA Home
  • Search Results

  • hydroquinone1101112331911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroquinone

8 Products
Synonym:   1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ
Image of Hydroquinone
CAS Number:
Linear Formula:
 C6H4-1,4-(OH)2
Molecular Weight:
 110.11
 Beilstein/REAXYS Number:
 605970
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Manufacturer designation
  Melting Point
  Anal. Standard Form
  pH Value
  Application(s)
  Solubility
  Product Line
  Physical Form
  Special Grade
Show More