• USA Home
  • Search Results

  • hydroxyapatite502311216774711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroxyapatite

5 Products
Synonym:   Calcium hydroxyphosphate, Calcium phosphate tribasic, HAp, Hydroxylapatite, Tribasic calcium phosphate
Image of Hydroxyapatite
CAS Number:
Linear Formula:
 [Ca5(OH)(PO4)3]x
Molecular Weight:
 502.31
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Additive
  Assay
  Surface Dimension
  Particle Dimension
Show More