• USA Home
  • Search Results

  • hydroxyapatite50231130606511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroxyapatite

3 Products
Synonym:   Calcium phosphate hydroxide, Durapatite, Hydroxylapatite
Image of Hydroxyapatite
CAS Number:
Linear Formula:
 3Ca3(PO4)2 · Ca(OH)2
Molecular Weight:
 502.31
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  Miscellaneous Specifications
  Loss
  Capacity
Show More