• USA Home
  • Search Results

  • hydroxycitronellal1722610775511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroxycitronellal

2 Products
Synonym:   7-Hydroxy-3,7-dimethyl-octanal
Image of Hydroxycitronellal
CAS Number:
Empirical Formula (Hill Notation):
 C10H20O2
Molecular Weight:
 172.26
 Council of Europe no.:
 100
 EC Number:
 FEMA Number:
 2583
 Flavis number:
 5.012
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Assay
  Special Grade
Show More