• USA Home
  • Search Results

  • hydroxypropylbetadex1234512844635511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hydroxypropyl betadex

2 Products
Synonym:   (2-Hydroxypropyl)-β-cyclodextrin
Image of Hydroxypropyl betadex
CAS Number:
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More