• USA Home
  • Search Results

  • hygromycinbfromstreptomyceshygroscopicus527523128204911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hygromycin B from Streptomyces hygroscopicus

3 Products
Image of Hygromycin B from <I>Streptomyces hygroscopicus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C20H37N3O13
Molecular Weight:
 527.52
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Suitability
  pK-Values
  Titertest
  Purification method
  Product Line
  Assay
  Mode of Action
  Physical Form
  Biological Source
  Activity Spectrum
  Solubility
Show More