• USA Home
  • Search Results

  • hygromycinbsolutionfromstreptomyceshygroscopicus527523128204911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hygromycin B solution from Streptomyces hygroscopicus

2 Products
Image of Hygromycin B solution from <I>Streptomyces hygroscopicus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C20H37N3O13
Molecular Weight:
 527.52
 Beilstein/REAXYS Number:
 6755837
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Mode of Action
  Activity Spectrum
  Sterilization
  Concentration
  Biological Source
  Assay
Show More