• USA Home
  • Search Results

  • hypericinfromhypericumperforatum5044454804911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hypericin from Hypericum perforatum

1 Products
Synonym:   4,5,7,4′,5′,7′-Hexahydroxy-2,2′-dimethylnaphthodianthrone, Cyclo werrol, Hypericum red
Image of Hypericin from <I>Hypericum perforatum</I>
CAS Number:
Empirical Formula (Hill Notation):
 C30H16O8
Molecular Weight:
 504.44
 Beilstein/REAXYS Number:
 1917913
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More