• USA Home
  • Search Results

  • hypromellose12345900465311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Hypromellose

3 Products
Synonym:   (Hydroxypropyl)methyl cellulose, Hydroxypropylmethylcellulose 2910
Image of Hypromellose
CAS Number:
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
Show More