• USA Home
  • Search Results

  • ibuprofen206281568727111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ibuprofen

7 Products
Synonym:   α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid
Image of Ibuprofen
CAS Number:
Empirical Formula (Hill Notation):
 C13H18O2
Molecular Weight:
 206.28
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Manufacturer designation
  Assay
  Anal. Standard Form
  Application(s)
  Physical Form
Show More