• USA Home
  • Search Results

  • ici204448hydrochloride5018312126404811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

ICI 204,448 hydrochloride

1 Products
Synonym:   (±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride, R,S-[3-[1-[[(3,4-Dichlorophenyl)acetyl]methylamino]-2-(1-pyrrolidinyl)ethyl] phenoxy]-acetic acid hydrochloride
Image of ICI 204,448 hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C23H26Cl2N2O4 · HCl
Molecular Weight:
 501.83
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More