• USA Home
  • Search Results

  • idaeinchloride484842766136511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Idaein chloride

2 Products
Synonym:   3-(Galactosyloxy)-3′,4′,5,7-tetrahydroxyflavylium chloride, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-galactoside chloride, Idein chloride
Image of Idaein chloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H21ClO11
Molecular Weight:
 484.84
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Application(s)
Show More