• USA Home
  • Search Results

  • idoxuridine354105442211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Idoxuridine

3 Products
Synonym:   1-(2-Deoxy-β-D-ribofuranosyl)-5-iodouracil, 2′-Deoxy-5-iodouridine, 5-IUdR, 5-Iodo-2′-deoxyuridine, IDU, IdUrd, Idoxuridine
Image of Idoxuridine
CAS Number:
Empirical Formula (Hill Notation):
 C9H11IN2O5
Molecular Weight:
 354.10
 Beilstein/REAXYS Number:
 30397
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More