• USA Home
  • Search Results

  • ifosfamide26109377873211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ifosfamide

4 Products
Synonym:   Ifex, N,3-Bis(2-chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide
Image of Ifosfamide
CAS Number:
Empirical Formula (Hill Notation):
 C7H15Cl2N2O2P
Molecular Weight:
 261.09
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Titertest
Show More