• USA Home
  • Search Results

  • imatinibmesylate5897122012757111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Imatinib mesylate

1 Products
Synonym:   4-[(4-Methyl-1-piperazinyl)methyl]-N-[4-methyl-3-[[4-(3-pyridinyl)-2-pyrimidinyl]amino]-phenyl]benzamide methanesulfonate, 4-[(4-Methylpiperazin-1-yl)methyl]-N-(4-methyl-3-{[4-(pyridin-3-yl)pyrimidin-2-yl]amino}phenyl)benzamide mesylate, CGP 57148, Genfatinib, Glivec, Imatinib methanesulfonate, STI-571
CAS Number:
Empirical Formula (Hill Notation):
 C29H31N7O · CH4O3S
Molecular Weight:
 589.71
 NACRES
 NA.77
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More