• USA Home
  • Search Results

  • imidazole680828832411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Imidazole

14 Products
Synonym:   Glyoxaline, Iminazole, Imidazole
CAS Number:
Empirical Formula (Hill Notation):
 C3H4N2
Molecular Weight:
 68.08
 EC Index Number:
 206-019-2
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  UV-Absorption Condition
  Purity Grade
  Product Line
  Potency
  Anal. Standard Form
  Greener Alternative Principles
  Vapor pressure
  Transition temperature
  Density
  Bulk Density
  Autoignition temp
  UV-Absorption Values
  Melting Point
  Solubility
  Physical Form
  pH Value
  Assay
Show More