• USA Home
  • Search Results

  • iminodiaceticacid1331014273411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iminodiacetic acid

3 Products
Synonym:   IDA
Image of Iminodiacetic acid
CAS Number:
Linear Formula:
 HN(CH2COOH)2
Molecular Weight:
 133.10
 Beilstein/REAXYS Number:
 878499
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
Show More