• USA Home
  • Search Results

  • imperatorin2702848244011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Imperatorin

3 Products
Synonym:   8-Isopentenyloxypsoralene, 9-(3-Methylbut-2-enyloxy)-7H-furo[3,2-g]chromen-7-one, 9-[(3-Methyl-2-buten-1-yl)oxy]-7h-furo[3,2-g][1]benzopyran-7-one, Ammidin, Marmelide, Marmelosin, NSC 402949
Image of Imperatorin
CAS Number:
Empirical Formula (Hill Notation):
 C16H14O4
Molecular Weight:
 270.28
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More