• USA Home
  • Search Results

  • indazole1181427144311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indazole

1 Products
Synonym:   1,2-Benzodiazole, 1,2-Benzopyrazole
Image of Indazole
CAS Number:
Empirical Formula (Hill Notation):
 C7H6N2
Molecular Weight:
 118.14
 Beilstein/REAXYS Number:
 109676
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
I2401 Indazole, 98%,