• USA Home
  • Search Results

  • indium11482744074611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indium

268 Products
Image of Indium
CAS Number:
Empirical Formula (Hill Notation):
 In
Molecular Weight:
 114.82
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Weight
  Particle Dimension
  Composite Dimension
  Manufacturer designation
  Physical Form
Show More

Product #
Description
277959 Indium, powder, 99.99% trace metals basis,
264032 Indium, powder, −100 mesh, 99.99% trace metals basis,
357308 Indium, foil, thickness 0.1 mm, ≥99.995% trace metals basis,
264113 Indium, beads, diam. 2-5 mm, 99.999% trace metals basis,
357073 Indium, wire, diam. 1.0 mm, 99.995% trace metals basis,
57083 Indium, bars, 99.95%,
326607 Indium, beads, diam. 2-5 mm, ≥99.9% trace metals basis,
264059 Indium, foil, thickness 0.127 mm, 99.99% trace metals basis,
203432 Indium, powder, −60 mesh, 99.999% trace metals basis,
264075 Indium, wire, diam. 1.0 mm, 99.99% trace metals basis,
264067 Indium, wire, diam. 0.5 mm, 99.995% trace metals basis,
264091 Indium, rod, diam. 6 mm, ≥99.999% trace metals basis,
357065 Indium, wire, diam. 2.0 mm, 99.995% trace metals basis,
GF97000860 Indium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.999%,
GF97195595 Indium, foil, not light tested, 50x50mm, thickness 0.0025mm, temporary acrylic support, 99.8%,
GF40799616 Indium, foil, 10mm disks, thickness 0.03mm, 99.8%,
GF74207347 Indium, wire reel, 1m, diameter 1.5mm, as drawn, 99.999%,
1.12196 Indium, granular 99+,
GF94338322 Indium, lump, 15 mm max. lump size, weight 100 g, high purity 99.999%,
GF98066043 Indium, lump, 8 mm max. lump size, high purity 99.999%, weight 50 g,
GF99101741 Indium, foil, thickness 0.15 mm, size 150 × 150 mm, high purity 99.999%,
GF62476656 Indium, rod, 2.0 mm diameter, length 100 mm, high purity 99.999%,
GF68616041 Indium, rod, 10.0 mm diameter, length 150 mm, high purity 99.999%,
GF74054606 Indium, lump, 15 mm max. lump size, weight 10 g, high purity 99.999%,
GF75395672 Indium, rod, length 500 mm, 2.0 mm diameter, high purity 99.999%,
GF77006033 Indium, rod, 10.0 mm diameter, length 300 mm, high purity 99.999%,
GF78519000 Indium, foil, thickness 0.15 mm, size 50 × 50 mm, high purity 99.999%,
GF80260837 Indium, rod, 3.5 mm diameter, length 100 mm, high purity 99.999%,
GF81807262 Indium, foil, thickness 0.15 mm, diameter 8 mm, high purity 99.999%,
GF83912924 Indium, foil, thickness 0.15 mm, diameter 25 mm, high purity 99.999%,
GF87981148 Indium, foil, thickness 0.15 mm, diameter 15 mm, high purity 99.999%,
GF90400104 Indium, foil, thickness 0.15 mm, diameter 4 mm, high purity 99.999%,
GF91171559 Indium, foil, thickness 0.15 mm, diameter 10 mm, high purity 99.999%,
GF91235077 Indium, rod, length 500 mm, 5.0 mm diameter, high purity 99.999%,
GF92290293 Indium, lump, 15 mm max. lump size, weight 200 g, high purity 99.999%,
GF94052538 Indium, rod, 3.5 mm diameter, length 1000 mm, high purity 99.999%,
GF00272797 Indium, rod, 5.0 mm diameter, length 200 mm, high purity 99.999%,
GF03154700 Indium, lump, 15 mm max. lump size, weight 50 g, high purity 99.999%,
GF04384807 Indium, rod, 5.0 mm diameter, length 1000 mm, high purity 99.999%,
GF05402810 Indium, rod, 3.5 mm diameter, length 200 mm, high purity 99.999%,
GF05777196 Indium, rod, 10.0 mm diameter, length 50 mm, high purity 99.999%,
GF09581429 Indium, rod, 2.0 mm diameter, length 200 mm, high purity 99.999%,
GF19202183 Indium, rod, 5.0 mm diameter, length 100 mm, high purity 99.999%,
GF22148928 Indium, rod, 10.0 mm diameter, length 100 mm, high purity 99.999%,
GF24458812 Indium, rod, 2.0 mm diameter, length 1000 mm, high purity 99.999%,
GF24572280 Indium, lump, 20 mm max. lump size, weight 40 g, high purity 99.99998%,
GF25809105 Indium, powder, max. particle size 150 micron, weight 10 g, high purity 99.999%,
GF26138268 Indium, powder, max. particle size 150 micron, weight 20 g, high purity 99.999%,
GF26725349 Indium, rod, 3.5 mm diameter, length 500 mm, high purity 99.999%,
GF34002751 Indium, lump, 8 mm max. lump size, weight 20 g, high purity 99.999%,
GF34099814 Indium, rod, 5.0 mm diameter, length 50 mm, high purity 99.999%,
GF35860776 Indium, lump, 20 mm max. lump size, weight 20 g, high purity 99.99998%,
GF36286881 Indium, lump, 20 mm max. lump size, weight 10 g, high purity 99.99999%,
GF39429546 Indium, lump, 8 mm max. lump size, weight 10 g, high purity 99.999%,
GF48100407 Indium, rod, 5.0 mm diameter, length 25 mm, high purity 99.99+%,
GF51743684 Indium, foil, thickness 0.15 mm, size 100 × 100 mm, high purity 99.999%,
GF55228428 Indium, foil, thickness 0.15 mm, 6 mm diameter, high purity 99.999%,
GF56825231 Indium, lump, 15 mm max. lump size, weight 20 g, high purity 99.999%,
GF58714308 Indium, foil, thickness 0.15 mm, size 25 × 25 mm, high purity 99.999%,
GF29634228 Indium, foil, 6mm disks, thickness 0.1mm, as rolled, 100%,
GF74341278 Indium, foil, 6mm disks, thickness 0.20mm, as rolled, 100%,
GF94704644 Indium, foil, 6mm disks, thickness 0.25mm, as rolled, 100%,
GF83950027 Indium, foil, 6mm disks, thickness 0.5mm, as rolled, 100%,
GF58678958 Indium, foil, 6mm disks, thickness 0.75mm, as rolled, 100%,
GF44006712 Indium, foil, 6mm disks, thickness 1.0mm, as rolled, 100%,
GF04295538 Indium, foil, 8mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF87572649 Indium, foil, 8mm disks, thickness 0.015mm, 99.8%,
GF85401918 Indium, foil, 8mm disks, thickness 0.01mm, 99.8%,
GF40183950 Indium, foil, 8mm disks, thickness 0.025mm, 99.8%,
GF89773138 Indium, foil, 8mm disks, thickness 0.02mm, 99.8%,
GF76326396 Indium, foil, 8mm disks, thickness 0.03mm, 99.8%,
GF06331047 Indium, foil, 8mm disks, thickness 0.04mm, 99.8%,
GF35535157 Indium, foil, 8mm disks, thickness 0.05mm, as rolled, 100%,
GF98837319 Indium, foil, 8mm disks, thickness 0.075mm, as rolled, 100%,
GF39812137 Indium, foil, 8mm disks, thickness 0.125mm, as rolled, 100%,
GF04277729 Indium, foil, 8mm disks, thickness 0.1mm, as rolled, 100%,
GF92659540 Indium, foil, 8mm disks, thickness 0.20mm, as rolled, 100%,
GF38080065 Indium, foil, 8mm disks, thickness 0.25mm, as rolled, 100%,
GF93752590 Indium, foil, 8mm disks, thickness 0.5mm, as rolled, 100%,
GF55033589 Indium, foil, 8mm disks, thickness 0.75mm, as rolled, 100%,
GF00838997 Indium, foil, 8mm disks, thickness 1.0mm, as rolled, 100%,
GF24434748 Indium, foil, light tested, 100x100mm, thickness 0.075mm, as rolled, 100%,
GF61338966 Indium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 100%,
GF29364981 Indium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 100%,
GF07860900 Indium, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 100%,
GF96094873 Indium, foil, not light tested, 100x100mm, thickness 0.006mm, temporary acrylic support, 99.8%,
GF97485823 Indium, foil, not light tested, 100x100mm, thickness 0.008mm, temporary acrylic support, 99.8%,
GF34180758 Indium, foil, not light tested, 100x100mm, thickness 0.009mm, temporary acrylic support, 99.8%,
GF21569373 Indium, foil, not light tested, 100x100mm, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF82494557 Indium, foil, not light tested, 100x100mm, thickness 0.015mm, 99.8%,
GF12014053 Indium, foil, not light tested, 100x100mm, thickness 0.01mm, 99.8%,
GF92066496 Indium, foil, not light tested, 100x100mm, thickness 0.025mm, 99.8%,
GF20336149 Indium, foil, not light tested, 100x100mm, thickness 0.02mm, 99.8%,
GF40478508 Indium, foil, not light tested, 100x100mm, thickness 0.03mm, 99.8%,
GF38498529 Indium, foil, not light tested, 100x100mm, thickness 0.04mm, 99.8%,
GF56607341 Indium, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.8%,
GF73195038 Indium, foil, not light tested, 25x25mm, thickness 0.002mm, temporary acrylic support, 99.8%,
GF53164650 Indium, foil, not light tested, 25x25mm, thickness 0.003mm, temporary acrylic support, 99.8%,
GF76403303 Indium, foil, not light tested, 25x25mm, thickness 0.004mm, temporary acrylic support, 99.8%,
GF11864932 Indium, foil, not light tested, 25x25mm, thickness 0.006mm, temporary acrylic support, 99.8%,
GF12678127 Indium, foil, not light tested, 25x25mm, thickness 0.008mm, temporary acrylic support, 99.8%,
GF97552941 Indium, foil, not light tested, 25x25mm, thickness 0.009mm, temporary acrylic support, 99.8%,
GF19313030 Indium, foil, not light tested, 25x25mm, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF44622086 Indium, foil, not light tested, 25x25mm, thickness 0.015mm, 99.8%,
GF50265918 Indium, foil, not light tested, 25x25mm, thickness 0.01mm, 99.8%,
GF28933858 Indium, foil, not light tested, 25x25mm, thickness 0.025mm, 99.8%,
GF37873738 Indium, foil, not light tested, 25x25mm, thickness 0.02mm, 99.8%,
GF78517042 Indium, foil, not light tested, 25x25mm, thickness 0.03mm, 99.8%,
GF22451420 Indium, foil, not light tested, 25x25mm, thickness 0.04mm, 99.8%,
GF88051114 Indium, foil, not light tested, 50x50mm, thickness 0.003mm, temporary acrylic support, 99.8%,
GF62925138 Indium, foil, not light tested, 50x50mm, thickness 0.004mm, temporary acrylic support, 99.8%,
GF79932123 Indium, foil, not light tested, 50x50mm, thickness 0.005mm, temporary acrylic support, 99.8%,
GF19585996 Indium, foil, not light tested, 50x50mm, thickness 0.006mm, temporary acrylic support, 99.8%,
GF99538600 Indium, foil, not light tested, 50x50mm, thickness 0.007mm, temporary acrylic support, 99.8%,
GF99016487 Indium, foil, not light tested, 50x50mm, thickness 0.008mm, temporary acrylic support, 99.8%,
GF70986864 Indium, foil, not light tested, 50x50mm, thickness 0.009mm, temporary acrylic support, 99.8%,
GF08164380 Indium, foil, not light tested, 50x50mm, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF10892404 Indium, foil, not light tested, 50x50mm, thickness 0.015mm, 99.8%,
GF87394652 Indium, foil, not light tested, 50x50mm, thickness 0.01mm, 99.8%,
GF08711052 Indium, foil, not light tested, 50x50mm, thickness 0.025mm, 99.8%,
GF41231797 Indium, foil, not light tested, 50x50mm, thickness 0.02mm, 99.8%,
GF50338436 Indium, foil, not light tested, 50x50mm, thickness 0.03mm, 99.8%,
GF69902084 Indium, foil, not light tested, 50x50mm, thickness 0.04mm, 99.8%,
GF15221348 Indium, wire reel, 0.1m, diameter 1.5mm, as drawn, 99.999%,
GF84663658 Indium, wire reel, 0.1m, diameter 1.8mm, as drawn, 99.999%,
GF97360779 Indium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.999%,
GF69686724 Indium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.999%,
GF63665488 Indium, wire reel, 0.2m, diameter 0.75mm, as drawn, 99.999%,
GF89275674 Indium, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.999%,
GF41790348 Indium, wire reel, 0.2m, diameter 1.5mm, as drawn, 99.999%,
GF25147278 Indium, wire reel, 0.2m, diameter 1.8mm, as drawn, 99.999%,
GF70097272 Indium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.999%,
GF63352087 Indium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%,
GF54155404 Indium, wire reel, 0.5m, diameter 0.75mm, as drawn, 99.999%,
GF59458911 Indium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%,
GF23793655 Indium, wire reel, 0.5m, diameter 1.5mm, as drawn, 99.999%,
GF26102821 Indium, wire reel, 0.5m, diameter 1.8mm, as drawn, 99.999%,
GF62158899 Indium, wire reel, 1m, diameter 0.25mm, as drawn, 99.999%,
GF19011068 Indium, wire reel, 1m, diameter 0.5mm, as drawn, 99.999%,
GF69425365 Indium, wire reel, 1m, diameter 0.75mm, as drawn, 99.999%,
GF32364536 Indium, wire reel, 1m, diameter 1.0mm, as drawn, 99.999%,
GF29580891 Indium, wire reel, 1m, diameter 1.8mm, as drawn, 99.999%,
GF69862382 Indium, wire reel, 2m, diameter 0.25mm, as drawn, 99.999%,
GF76383254 Indium, wire reel, 2m, diameter 0.5mm, as drawn, 99.999%,
GF78170003 Indium, wire reel, 2m, diameter 0.75mm, as drawn, 99.999%,
GF45566433 Indium, wire reel, 2m, diameter 1.0mm, as drawn, 99.999%,
GF75918099 Indium, wire reel, 2m, diameter 1.5mm, as drawn, 99.999%,
GF14314014 Indium, wire reel, 2m, diameter 1.8mm, as drawn, 99.999%,
GF32797659 Indium, wire reel, 5m, diameter 0.25mm, as drawn, 99.999%,
GF47907467 Indium, wire reel, 5m, diameter 0.5mm, as drawn, 99.999%,
GF71250511 Indium, wire reel, 5m, diameter 0.75mm, as drawn, 99.999%,
GF95804275 Indium, wire reel, 5m, diameter 1.0mm, as drawn, 99.999%,
GF89761652 Indium, wire reel, 5m, diameter 1.5mm, as drawn, 99.999%,
GF38467301 Indium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.999%,
GF04865564 Indium, foil, 100x100mm, thickness 0.1mm, as rolled, 100%,
GF40076276 Indium, foil, 100x100mm, thickness 0.20mm, as rolled, 99.999%,
GF25107690 Indium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.999%,
GF31489915 Indium, foil, 100x100mm, thickness 0.5mm, as rolled, 99.999%,
GF49523081 Indium, foil, 100x100mm, thickness 0.75mm, as rolled, 99.999%,
GF29431694 Indium, foil, 100x100mm, thickness 2.0mm, as rolled, 99.999%,
GF61355617 Indium, foil, 100x100mm, thickness 3.0mm, as rolled, 99.999%,
GF24254704 Indium, foil, 100x200mm, thickness 0.5mm, as rolled, 99.999%,
GF08413400 Indium, foil, 150x150mm, thickness 0.125mm, as rolled, 99.999%,
GF08195294 Indium, foil, 150x150mm, thickness 0.1mm, as rolled, 100%,
GF09534359 Indium, foil, 150x150mm, thickness 0.20mm, as rolled, 99.999%,
GF38461338 Indium, foil, 150x150mm, thickness 0.25mm, as rolled, 99.999%,
GF56337992 Indium, foil, 150x150mm, thickness 0.75mm, as rolled, 99.999%,
GF70592586 Indium, foil, 150x150mm, thickness 1.0mm, as rolled, 99.999%,
GF47151266 Indium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.999%,
GF30611517 Indium, foil, 25x25mm, thickness 0.1mm, as rolled, 100%,
GF02639369 Indium, foil, 25x25mm, thickness 0.20mm, as rolled, 99.999%,
GF83176005 Indium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.999%,
GF28328418 Indium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%,
GF91615068 Indium, foil, 25x25mm, thickness 0.75mm, as rolled, 99.999%,
GF84655198 Indium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%,
GF92065888 Indium, foil, 25x25mm, thickness 2.0mm, as rolled, 99.999%,
GF29953402 Indium, foil, 25x25mm, thickness 3.0mm, as rolled, 99.999%,
GF85273577 Indium, foil, 50x100mm, thickness 0.1mm, as rolled, 100%,
GF93907738 Indium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.999%,
GF26438924 Indium, foil, 50x50mm, thickness 0.1mm, as rolled, 100%,
GF29920619 Indium, foil, 50x50mm, thickness 0.20mm, as rolled, 99.999%,
GF41051911 Indium, foil, 50x50mm, thickness 0.5mm, as rolled, 99.999%,
GF40139658 Indium, foil, 50x50mm, thickness 0.75mm, as rolled, 99.999%,
GF81898914 Indium, foil, 50x50mm, thickness 1.0mm, as rolled, 99.999%,
GF08235457 Indium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.999%,
GF03993868 Indium, foil, 50x50mm, thickness 3.0mm, as rolled, 99.999%,
GF37247609 Indium, foil, not light tested, 150x150mm, thickness 0.075mm, as rolled, 100%,
GF79077998 Indium, foil, not light tested, 25x25mm, thickness 0.05mm, as rolled, 100%,
GF26682396 Indium, foil, not light tested, 75x100mm, thickness 0.05mm, as rolled, 100%,
GF97860549 Indium, foil, not light tested, 75x150mm, thickness 0.05mm, as rolled, 100%,
GF42899147 Indium, foil, not light tested, 75x300mm, thickness 0.05mm, as rolled, 100%,
GF36942419 Indium, foil, 10mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF46580957 Indium, foil, 10mm disks, thickness 0.015mm, 99.8%,
GF75239781 Indium, foil, 10mm disks, thickness 0.01mm, 99.8%,
GF22994578 Indium, foil, 10mm disks, thickness 0.025mm, 99.8%,
GF21641453 Indium, foil, 10mm disks, thickness 0.02mm, 99.8%,
GF91381204 Indium, foil, 10mm disks, thickness 0.04mm, 99.8%,
GF41894062 Indium, foil, 10mm disks, thickness 0.05mm, as rolled, 100%,
GF54664522 Indium, foil, 10mm disks, thickness 0.075mm, as rolled, 100%,
GF76516364 Indium, foil, 10mm disks, thickness 0.125mm, as rolled, 100%,
GF34716180 Indium, foil, 10mm disks, thickness 0.1mm, as rolled, 100%,
GF82443225 Indium, foil, 10mm disks, thickness 0.20mm, as rolled, 100%,
GF60008486 Indium, foil, 10mm disks, thickness 0.25mm, as rolled, 100%,
GF19074933 Indium, foil, 10mm disks, thickness 0.5mm, as rolled, 100%,
GF00134288 Indium, foil, 10mm disks, thickness 0.75mm, as rolled, 100%,
GF04874351 Indium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%,
GF17802244 Indium, foil, 15mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF22619101 Indium, foil, 15mm disks, thickness 0.015mm, 99.8%,
GF04972563 Indium, foil, 15mm disks, thickness 0.01mm, 99.8%,
GF74321894 Indium, foil, 15mm disks, thickness 0.025mm, 99.8%,
GF05181814 Indium, foil, 15mm disks, thickness 0.02mm, 99.8%,
GF99951610 Indium, foil, 15mm disks, thickness 0.03mm, 99.8%,
GF22650645 Indium, foil, 15mm disks, thickness 0.04mm, 99.8%,
GF36757660 Indium, foil, 15mm disks, thickness 0.05mm, as rolled, 100%,
GF33065232 Indium, foil, 15mm disks, thickness 0.075mm, as rolled, 100%,
GF64546106 Indium, foil, 15mm disks, thickness 0.125mm, as rolled, 100%,
GF39523091 Indium, foil, 15mm disks, thickness 0.1mm, as rolled, 100%,
GF05403439 Indium, foil, 15mm disks, thickness 0.20mm, as rolled, 100%,
GF01060366 Indium, foil, 15mm disks, thickness 0.25mm, as rolled, 100%,
GF84367301 Indium, foil, 15mm disks, thickness 0.5mm, as rolled, 100%,
GF24476227 Indium, foil, 15mm disks, thickness 0.75mm, as rolled, 100%,
GF09294992 Indium, foil, 15mm disks, thickness 1.0mm, as rolled, 100%,
GF33153959 Indium, foil, 25mm disks, thickness 0.075mm, as rolled, 100%,
GF68262565 Indium, foil, 25mm disks, thickness 0.125mm, as rolled, 100%,
GF40387013 Indium, foil, 25mm disks, thickness 0.1mm, as rolled, 100%,
GF75429322 Indium, foil, 25mm disks, thickness 0.20mm, as rolled, 100%,
GF32545818 Indium, foil, 25mm disks, thickness 0.25mm, as rolled, 100%,
GF45912618 Indium, foil, 25mm disks, thickness 0.5mm, as rolled, 100%,
GF66110060 Indium, foil, 25mm disks, thickness 0.75mm, as rolled, 100%,
GF61874691 Indium, foil, 25mm disks, thickness 1.0mm, as rolled, 100%,
GF27334672 Indium, foil, 4mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF57106403 Indium, foil, 4mm disks, thickness 0.015mm, 99.8%,
GF38968666 Indium, foil, 4mm disks, thickness 0.01mm, 99.8%,
GF29602571 Indium, foil, 4mm disks, thickness 0.025mm, 99.8%,
GF51442545 Indium, foil, 4mm disks, thickness 0.02mm, 99.8%,
GF49944088 Indium, foil, 4mm disks, thickness 0.03mm, 99.8%,
GF83085206 Indium, foil, 4mm disks, thickness 0.04mm, 99.8%,
GF22114279 Indium, foil, 4mm disks, thickness 0.05mm, as rolled, 100%,
GF62636722 Indium, foil, 4mm disks, thickness 0.075mm, as rolled, 100%,
GF51676140 Indium, foil, 4mm disks, thickness 0.125mm, as rolled, 100%,
GF51437077 Indium, foil, 4mm disks, thickness 0.1mm, as rolled, 100%,
GF91794730 Indium, foil, 4mm disks, thickness 0.20mm, as rolled, 100%,
GF58357637 Indium, foil, 4mm disks, thickness 0.25mm, as rolled, 100%,
GF43871609 Indium, foil, 4mm disks, thickness 0.5mm, as rolled, 100%,
GF28742890 Indium, foil, 4mm disks, thickness 0.75mm, as rolled, 100%,
GF09152052 Indium, foil, 4mm disks, thickness 1.0mm, as rolled, 100%,
GF34336526 Indium, foil, 50mm disks, thickness 1.0mm, as rolled, 100%,
GF16834361 Indium, foil, 6mm disks, thickness 0.0125mm, temporary acrylic support, 99.8%,
GF38300587 Indium, foil, 6mm disks, thickness 0.015mm, 99.8%,
GF71083212 Indium, foil, 6mm disks, thickness 0.01mm, 99.8%,
GF00321477 Indium, foil, 6mm disks, thickness 0.025mm, 99.8%,
GF30318184 Indium, foil, 6mm disks, thickness 0.02mm, 99.8%,
GF43010615 Indium, foil, 6mm disks, thickness 0.03mm, 99.8%,
GF85650644 Indium, foil, 6mm disks, thickness 0.04mm, 99.8%,
GF64284284 Indium, foil, 6mm disks, thickness 0.05mm, as rolled, 100%,
GF29590690 Indium, foil, 6mm disks, thickness 0.075mm, as rolled, 100%,
GF59518491 Indium, foil, 6mm disks, thickness 0.125mm, as rolled, 100%,
GF47226191 Indium, foil, not light tested, 25x25mm, thickness 0.007mm, temporary acrylic support, 99.8%,
GF84050333 Indium, foil, not light tested, 50x50mm, thickness 0.002mm, temporary acrylic support, 99.8%,
GF62573395 Indium, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 100%,
264040 Indium, foil, thickness 0.25 mm, 99.995% trace metals basis,
326615 Indium, pieces, 99.99% trace metals basis,
326623 Indium, wire, diam. 0.25 mm, 99.999% trace metals basis,
326631 Indium, foil, thickness 0.5 mm, 99.999% trace metals basis,
357278 Indium, foil, thickness 1.0 mm, 99.999% trace metals basis,
357294 Indium, foil, thickness 0.25 mm, 99.99% trace metals basis,
357286 Indium, foil, thickness 0.5 mm, 99.99% trace metals basis,
340863 Indium, wire, diam. 1.5 mm, 99.99% trace metals basis,