• USA Home
  • Search Results

  • indole1171512072911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indole

5 Products
Synonym:   1H-Benzo[b]pyrrole
Image of Indole
CAS Number:
Empirical Formula (Hill Notation):
 C8H7N
Molecular Weight:
 117.15
 Beilstein/REAXYS Number:
 107693
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Potency
  Melting Point
  Biological Source
  Agency/Method
  Special Grade
  Assay
Show More