• USA Home
  • Search Results

  • indole2carboxylicacid16116147750511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indole-2-carboxylic acid

1 Products
Synonym:   2-Carboxyindole, 2-Indolylformic acid, NSC 16598
Image of Indole-2-carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C9H7NO2
Molecular Weight:
 161.16
 Beilstein/REAXYS Number:
 124132
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More