• USA Home
  • Search Results

  • indole4carboxaldehyde14516107486811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indole-4-carboxaldehyde

1 Products
Synonym:   4-Formylindole, NSC 337264
Image of Indole-<WBR>4-<WBR>carboxaldehyde
CAS Number:
Empirical Formula (Hill Notation):
 C9H7NO
Molecular Weight:
 145.16
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More