• USA Home
  • Search Results

  • indole5carbonitrile142161586124211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indole-5-carbonitrile

1 Products
Synonym:   5-Cyanoindole
Image of Indole-<WBR>5-<WBR>carbonitrile
CAS Number:
Empirical Formula (Hill Notation):
 C9H6N2
Molecular Weight:
 142.16
 Beilstein/REAXYS Number:
 116738
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More