• USA Home
  • Search Results

  • indomethacin357795386111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Indomethacin

5 Products
Synonym:   1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-3-indoleacetic acid
Image of Indomethacin
CAS Number:
Empirical Formula (Hill Notation):
 C19H16ClNO4
Molecular Weight:
 357.79
 Beilstein/REAXYS Number:
 497341
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Melting Point
  Mode of Action
  Anal. Standard Form
  Special Grade
  Assay
Show More