• USA Home
  • Search Results

  • inositol180168789811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Inositol

2 Products
Synonym:   1,2,3,4,5,6-Hexahydroxycyclohexane, i-Inositol, meso-Inositol, myo-Inositol
Image of Inositol
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1907329
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More