• USA Home
  • Search Results

  • insulinhuman5807571106168011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Insulin human

7 Products
CAS Number:
Empirical Formula (Hill Notation):
 C257H383N65O77S6
Molecular Weight:
 5807.57
 EC Number:
 MDL number:
 NACRES:
 NA.32
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Sterilization
  Recombinant Host
  Potency
  Manufacturer designation
  Biological Source
  Solubility
  Ice Shipping
  Assay
  Application(s)
Show More