• USA Home
  • Search Results

  • iodicacid17591778268511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iodic acid

2 Products
Image of Iodic acid
CAS Number:
Empirical Formula (Hill Notation):
 HIO3
Molecular Weight:
 175.91
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
 eCl@ss
 32110203
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
  Solubility
  Density
Show More