• USA Home
  • Search Results

  • iodomethyltrimethylsilane21412420667111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Iodomethyl)trimethylsilane

1 Products
Synonym:   (Trimethylsilyl)methyl iodide
Image of (Iodomethyl)<WBR>trimethylsilane
CAS Number:
Linear Formula:
 (CH3)3SiCH2I
Molecular Weight:
 214.12
 Beilstein/REAXYS Number:
 1731485
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More