• USA Home
  • Search Results

  • iodosulfuronmethylsodium5292414455036711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iodosulfuron-methyl-sodium

1 Products
Synonym:   Methyl 4-iodo-2-[3-(4-methoxy-6-methyl-1,3,5-triazin-2-yl)ureidosulfonyl]benzoate sodium salt
Image of Iodosulfuron-<WBR>methyl-<WBR>sodium
CAS Number:
Empirical Formula (Hill Notation):
 C14H13IN5NaO6S
Molecular Weight:
 529.24
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More