• USA Home
  • Search Results

  • ionnylonabsalloy3dprintingfilament12345900356911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

iON nylon/ABS alloy 3D printing filament

3 Products
CAS Number:
 NACRES
 NA.23
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Color
  Composite Dimension
Show More