• USA Home
  • Search Results

  • ionomycinfromstreptomycesconglobatus709005609281011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ionomycin from Streptomyces conglobatus

1 Products
Image of Ionomycin from <I>Streptomyces conglobatus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C41H72O9
Molecular Weight:
 709.00
 Beilstein/REAXYS Number:
 3642126
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More