• USA Home
  • Search Results

  • ir10617491315561401011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

IR-1061

1 Products
Synonym:   4-[2-[2-Chloro-3-[(2,6-diphenyl-4H-thiopyran-4-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-2,6-diphenylthiopyrylium tetrafluoroborate
Image of IR-1061
CAS Number:
Empirical Formula (Hill Notation):
 C44H34ClS2 · BF4
Molecular Weight:
 749.13
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More