• USA Home
  • Search Results

  • ir797chloride5055211099255711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

IR-797 chloride

1 Products
Synonym:   2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,3,3-trimethyl-2H-indol-2-ylidene)-ethylidene]-1-cyclopenten-1-yl]-ethenyl]-1,3,3-trimethyl-3H-indolium chloride
Image of IR-797 chloride
CAS Number:
Empirical Formula (Hill Notation):
 C31H34Cl2N2
Molecular Weight:
 505.52
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More