• USA Home
  • Search Results

  • irdfcf3ppy2dtbpypf611219187098763611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(Ir[dF(CF3)ppy]2(dtbpy))PF6

1 Products
Synonym:   [4,4′-Bis(1,1-dimethylethyl)-2,2′-bipyridine-N1,N1′]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-N]phenyl-C]Iridium(III) hexafluorophosphate, [Ir{dF(CF3)ppy}2(dtbpy)]PF6
Image of (Ir[dF(CF<SUB>3</SUB>)ppy]<SUB>2</SUB>(dtbpy))PF<SUB>6</SUB>
CAS Number:
Empirical Formula (Hill Notation):
 C42H34F16IrN4P
Molecular Weight:
 1121.91
 NACRES:
 NA.22
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More