• USA Home
  • Search Results

  • iridium19222743988511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iridium

98 Products
Image of Iridium
CAS Number:
Empirical Formula (Hill Notation):
 Ir
Molecular Weight:
 192.22
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Weight
  Particle Dimension
  Composite Dimension
  Assay
  Physical Form
Show More

Product #
Description
209686 Iridium, powder, 99.9% trace metals basis,
449229 Iridium, evaporation slug, diam. × L 0.6 cm × 1.2 cm, 99.9% trace metals basis,
336793 Iridium, black, powder, −200 mesh, ≥99% trace metals basis,
357324 Iridium, foil, thickness 0.25 mm, 99.9% trace metals basis,
266841 Iridium, wire, diam. 0.15 mm, 99.9% trace metals basis,
GF35321172 Iridium, rod, 50mm, diameter 3.0mm, as drawn, 99.9%,
GF07750795 Iridium, wire reel, 0.025m, diameter 2.0mm, as drawn, 99.9%,
GF44052667 Iridium, wire reel, 0.05m, diameter 0.075mm, as drawn, 99.9%,
GF97286428 Iridium, wire reel, 0.05m, diameter 0.1mm, as drawn, 99.9%,
GF22425375 Iridium, wire reel, 0.05m, diameter 0.8mm, as drawn, 99.9%,
GF68849544 Iridium, wire reel, 0.05m, diameter 2.0mm, as drawn, 99.9%,
GF06583394 Iridium, wire reel, 0.1m, diameter 0.075mm, as drawn, 99.9%,
GF65585021 Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.5%,
GF88210447 Iridium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9%,
GF26409247 Iridium, wire reel, 0.1m, diameter 0.15mm, as drawn, 99.9%,
GF60803412 Iridium, wire reel, 0.1m, diameter 0.1mm, as drawn, 99.9%,
GF31863607 Iridium, wire reel, 0.1m, diameter 0.25mm, as drawn, 99.9%,
GF99945534 Iridium, wire reel, 0.1m, diameter 0.2mm, as drawn, 99.9%,
GF36309450 Iridium, wire reel, 0.1m, diameter 0.38mm, as drawn, 99.9%,
GF50407062 Iridium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9%,
GF80911403 Iridium, wire reel, 0.1m, diameter 0.8mm, as drawn, 99.9%,
GF22923974 Iridium, wire reel, 0.2m, diameter 0.075mm, as drawn, 99.9%,
GF30370924 Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.5%,
GF26305130 Iridium, wire reel, 0.2m, diameter 0.125mm, as drawn, 99.9%,
GF01155170 Iridium, wire reel, 0.2m, diameter 0.15mm, as drawn, 99.9%,
GF00427250 Iridium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.9%,
GF82950273 Iridium, wire reel, 0.2m, diameter 0.25mm, as drawn, 99.9%,
GF60570176 Iridium, wire reel, 0.2m, diameter 0.2mm, as drawn, 99.9%,
GF20836425 Iridium, foil, 0.025m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%,
GF10109629 Iridium, rod, 25mm, diameter 3.0mm, as drawn, 99.9%,
GF00604853 Iridium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%,
GF20804004 Iridium, wire reel, 0.1m, diameter 0.177mm, as drawn, 99.9%,
GF75606992 Iridium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9%,
GF45723764 Iridium, wire reel, 0.2m, diameter 0.177mm, as drawn, 99.9%,
GF48997478 Iridium, wire reel, 0.2m, diameter 0.38mm, as drawn, 99.9%,
GF00883739 Iridium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.9%,
GF36739277 Iridium, wire reel, 0.2m, diameter 0.8mm, as drawn, 99.9%,
GF50874093 Iridium, wire reel, 0.2m, diameter 1.0mm, as drawn, 99.9%,
GF10466760 Iridium, wire reel, 0.3m, diameter 0.5mm, as drawn, 99.9%,
GF08345639 Iridium, wire reel, 0.4m, diameter 0.5mm, as drawn, 99.9%,
GF65497427 Iridium, wire reel, 0.5m, diameter 0.075mm, as drawn, 99.9%,
GF01838101 Iridium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.5%,
GF88141990 Iridium, wire reel, 0.5m, diameter 0.125mm, as drawn, 99.9%,
GF44396703 Iridium, wire reel, 0.5m, diameter 0.15mm, as drawn, 99.9%,
GF81708644 Iridium, wire reel, 0.5m, diameter 0.177mm, as drawn, 99.9%,
GF53407245 Iridium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.9%,
GF68784025 Iridium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9%,
GF71700432 Iridium, wire reel, 0.5m, diameter 0.2mm, as drawn, 99.9%,
GF25136490 Iridium, wire reel, 0.5m, diameter 0.38mm, as drawn, 99.9%,
GF43204834 Iridium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9%,
GF87178292 Iridium, wire reel, 1m, diameter 0.075mm, as drawn, 99.9%,
GF06236063 Iridium, wire reel, 1m, diameter 0.125mm, as drawn, 99.5%,
GF17748894 Iridium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9%,
GF49588138 Iridium, wire reel, 1m, diameter 0.15mm, as drawn, 99.9%,
GF19502164 Iridium, wire reel, 1m, diameter 0.177mm, as drawn, 99.9%,
GF75905680 Iridium, wire reel, 1m, diameter 0.1mm, as drawn, 99.9%,
GF98003257 Iridium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9%,
GF92270493 Iridium, wire reel, 1m, diameter 0.2mm, as drawn, 99.9%,
GF63461303 Iridium, wire reel, 2m, diameter 0.125mm, as drawn, 99.5%,
GF18611276 Iridium, wire reel, 2m, diameter 0.125mm, as drawn, 99.9%,
GF98517235 Iridium, wire reel, 2m, diameter 0.15mm, as drawn, 99.9%,
GF26117490 Iridium, wire, straight, 100mm, diameter 0.5mm, as drawn, 99.9%,
GF99450254 Iridium, wire, straight, 100mm, diameter 0.6mm, as drawn, 99.9%,
GF04592730 Iridium, wire, straight, 100mm, diameter 0.7mm, as drawn, 99.9%,
GF50717156 Iridium, wire, straight, 100mm, diameter 0.8mm, as drawn, 99.9%,
GF17010986 Iridium, wire, straight, 100mm, diameter 1.0mm, as drawn, 99.9%,
GF55206930 Iridium, wire, straight, 200mm, diameter 0.5mm, as drawn, 99.9%,
GF33689065 Iridium, wire, straight, 200mm, diameter 0.6mm, as drawn, 99.9%,
GF43612556 Iridium, wire, straight, 200mm, diameter 0.7mm, as drawn, 99.9%,
GF43690091 Iridium, wire, straight, 200mm, diameter 0.8mm, as drawn, 99.9%,
GF50335433 Iridium, wire, straight, 200mm, diameter 1.0mm, as drawn, 99.9%,
GF97994468 Iridium, wire, straight, 500mm, diameter 0.5mm, as drawn, 99.9%,
GF79441331 Iridium, wire, straight, 50mm, diameter 0.6mm, as drawn, 99.9%,
GF88166627 Iridium, wire, straight, 50mm, diameter 0.7mm, as drawn, 99.9%,
GF08427292 Iridium, wire, straight, 50mm, diameter 0.8mm, as drawn, 99.9%,
GF05163904 Iridium, wire, straight, 50mm, diameter 1.0mm, as drawn, 99.9%,
GF96543209 Iridium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%,
GF98486982 Iridium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%,
GF21596160 Iridium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%,
GF81468030 Iridium, foil, 0.05m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%,
GF81040463 Iridium, foil, 0.18m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%,
GF15807900 Iridium, foil, 0.1m coil, thickness 0.05mm, coil width .5mm, as rolled, 99.9%,
GF35631200 Iridium, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.9%,
GF07858974 Iridium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%,
GF63698305 Iridium, microfoil, disks, 10mm, thinness 0.1μm, specific density 224μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF86062923 Iridium, microfoil, disks, 25mm, thinness 0.1μm, specific density 224μg/cm2, permanent mylar 3.5μm support, 99.9%,
GF51827328 Iridium, rod, 25mm, diameter 2.0mm, as drawn, 99.9%,
GF92005220 Iridium, rod, 50mm, diameter 2.0mm, as drawn, 99.9%,
GF00803393 Iridium, wire, 0.25 mm diameter, size 100 mm, purity 99.9%,
GF12220467 Iridium, rod, 4.0 mm diameter, length 25 mm, purity 99.9%,
GF33912985 Iridium, powder, max. particle size 50 micron, weight 1 g, purity 99.9%,
GF39123558 Iridium, powder, max. particle size 50 micron, weight 2 g, purity 99.9%,
GF63430949 Iridium, wire, 0.25 mm diameter, size 1000 mm, purity 99.9%,
GF67173127 Iridium, wire, 0.25 mm diameter, size 500 mm, purity 99.9%,
GF74116728 Iridium, powder, 850 max. part. size (micron), high purity 99.995%, weight 1 g,
GF82934703 Iridium, foil, thickness 0.1 mm, size 25 × 25 mm, purity 99.9%,
GF95951111 Iridium, wire, 0.25 mm diameter, size 200 mm, purity 99.9%,
GF96473234 Iridium, foil, thickness 0.03 mm, size 25 × 25 mm, purity 99.9%,