• USA Home
  • Search Results

  • iridiumiiichloridehydrate29858anhydrousbasis1499661311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iridium(III) chloride hydrate

2 Products
Synonym:   IrCl3, Iridium trichloride hydrate
Image of Iridium(III) chloride hydrate
CAS Number:
Linear Formula:
 IrCl3 · xH2O
Molecular Weight:
 298.58 (anhydrous basis)
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More