• USA Home
  • Search Results

  • ironiiichloridehexahydrate270301002577111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron(III) chloride hexahydrate

5 Products
Synonym:   Ferric chloride hexahydrate
Image of Iron(III) chloride hexahydrate
CAS Number:
Linear Formula:
 FeCl3 · 6H2O
Molecular Weight:
 270.30
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Agency/Method
  pH Value
  Potency
  Bulk Density
  Purity Grade
  Special Grade
Show More