• USA Home
  • Search Results

  • ironiiinitratenonahydrate40400778261811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron(III) nitrate nonahydrate

5 Products
Synonym:   Ferric nitrate nonahydrate
Image of Iron(III) nitrate nonahydrate
CAS Number:
Linear Formula:
 Fe(NO3)3 · 9H2O
Molecular Weight:
 404.00
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Special Grade
Show More