• USA Home
  • Search Results

  • ironiiioxidedispersion15969130937111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron(III) oxide, dispersion

1 Products
Synonym:   Iron oxide in ethanol
Image of Iron(III) oxide, dispersion
CAS Number:
Empirical Formula (Hill Notation):
 Fe2O3
Molecular Weight:
 159.69
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Particle Dimension
Show More