• USA Home
  • Search Results

  • ironiioxalatedihydrate17989604725211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron(II) oxalate dihydrate

2 Products
Synonym:   Ferrous oxalate dihydrate
Image of Iron(II) oxalate dihydrate
CAS Number:
Linear Formula:
 FeC2O4 · 2H2O
Molecular Weight:
 179.89
 Beilstein/REAXYS Number:
 3757620
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
Show More