• USA Home
  • Search Results

  • ironiisulfateheptahydrate27801778263011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron(II) sulfate heptahydrate

7 Products
Synonym:   Ferrous sulfate heptahydrate
Image of Iron(II) sulfate heptahydrate
CAS Number:
Linear Formula:
 FeSO4 · 7H2O
Molecular Weight:
 278.01
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Shelf Life
  Purity Grade
  Potency
  Melting Point
  Density
  Bulk Density
  Application(s)
  Reaction Suitability
  Product Line
  Special Grade
  Agency/Method
  Physical Form
  pH Value
Show More