• USA Home
  • Search Results

  • ironoxideincorporatedconjugatedpolymernanoparticles1234598765

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Iron oxide incorporated conjugated polymer nanoparticles

7 Products
Synonym:   Conjugated polymer nanoparticles, Fluorescent magnetic nanoparticles, Fluorescent nanoparticles, Magneto-conjugated polymer nanoparticles
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Physical Form
  Fluorescence Test
Show More

Product #
Description
904996 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 680 nm, 100 μg/mL in H2O,
905054 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 475 nm, 100 μg/mL in H2O,
905046 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 550 nm, 100 μg/mL in H2O,
913960 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 420 nm, 100 μg/mL in H2O, New
914215 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 610 nm, 100 μg/mL in H2O, New
913707 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 435 nm, 100 μg/mL in H2O, New
905038 Iron oxide incorporated conjugated polymer nanoparticles, fluorescence λem 510 nm, 100 μg/mL in H2O,