• USA Home
  • Search Results

  • irsogladinemaleate372168450469811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Irsogladine maleate

1 Products
Synonym:   2,4-Diamino-6-(2,5-dichlorophenyl)-s-triazine maleate, 6-(2,5-Dichlorophenyl)-1,3,5-triazine-2,4-diamine (2Z)-2-butenedioate (1:1), Gaslon N, MN 1695
Image of Irsogladine maleate
CAS Number:
Empirical Formula (Hill Notation):
 C9H7Cl2N5 ·C4H4O4
Molecular Weight:
 372.16
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More