• USA Home
  • Search Results

  • isatin147139156511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isatin

2 Products
Synonym:   2,3-Dioxo-2,3-dihydroindole, 2,3-Diketoindoline, 2,3-Dioxoindoline, 2,3-Indolinedione, o-Aminobenzoylformic anhydride, Isatic acid lactam, Isatine, Isatinic acid anhydride, NSC 9262
Image of Isatin
CAS Number:
Empirical Formula (Hill Notation):
 C8H5NO2
Molecular Weight:
 147.13
 Beilstein/REAXYS Number:
 383659
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Melting Point
  Assay
Show More