• USA Home
  • Search Results

  • isoamylacetate1301812392211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoamyl acetate

3 Products
Synonym:   Acetic acid 3-methylbutyl ester, Isoamyl acetate, Isopentyl acetate
Image of Isoamyl acetate
CAS Number:
Linear Formula:
 CH3COOCH2CH2CH(CH3)2
Molecular Weight:
 130.18
 Beilstein/REAXYS Number:
 1744750
 Council of Europe no.:
 214
 EC Number:
 FEMA Number:
 2055
 Flavis number:
 9.024
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Potency
  Assay
  Special Grade
Show More