• USA Home
  • Search Results

  • isoamylalcohol881512351311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Isoamyl alcohol

5 Products
Synonym:   3-Methyl-1-butanol, Isoamyl alcohol, Isopentyl alcohol
Image of Isoamyl alcohol
CAS Number:
Linear Formula:
 (CH3)2CHCH2CH2OH
Molecular Weight:
 88.15
 Beilstein/REAXYS Number:
 1718835
 Council of Europe no.:
 51
 EC Number:
 FEMA Number:
 2057
 Flavis number:
 2.003
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Allergen
  Organoleptic property
  Viscosity
  Agency/Method
  Special Grade
  Transition temperature
  Solubility
  Potency
  Assay
  Physical Form
  pH Value
Show More